missing translation for 'onlineSavingsMsg'
Learn More

2-(N-Methyl-2-aminoethyl)-1,3-dioxolane 98.0+%, TCI America™

Catalog No. M10525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10525G Supplier TCI America Supplier No. M10525G
Only null left

Chemical Identifiers

CAS 142753-10-4
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00191459
InChI Key ZUUQKDDUUOFCNP-UHFFFAOYSA-N
PubChem CID 21975567
IUPAC Name [2-(1,3-dioxolan-2-yl)ethyl](methyl)amine
SMILES CNCCC1OCCO1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 131.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(N-Methyl-2-aminoethyl)-1,3-dioxolane