missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexyloxybenzaldehyde, 96%, Thermo Scientific Chemicals

Catalog No. AAL0898809
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0898809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0898809 Supplier Thermo Scientific Chemicals Supplier No. L0898809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7162-59-6 | Molecular Formula: C13H18O2 | Molecular Weight: 206.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7162-59-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00014131
InChI Key IFOIDROUJIGQAV-UHFFFAOYSA-N
Synonym o-hexyloxybenzaldehyde, 2-hexyloxy benzaldehyde, 2-n-hexyloxybenzaldehyde, 2-hexyloxybenzaldehyde, hexyloxybenzaldehyde, benzaldehyde, 2-hexyloxy, benzaldehyde, o-n-hexyloxy-,
PubChem CID 138951
IUPAC Name 2-hexoxybenzaldehyde
SMILES CCCCCCOC1=CC=CC=C1C=O

Specifications

Boiling Point 132°C to 134°C (0.5 mmHg)
Refractive Index 1.52
Quantity 10 g
Beilstein 2258613
Sensitivity Air Sensitive
Formula Weight 206.29
Percent Purity 96%
Chemical Name or Material 2-n-Hexyloxybenzaldehyde
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only