missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexylfuran, 97%

Catalog No. p-7029253
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2669803 1 g
AAH2669806 5 g
AAH2669814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2669803 Supplier Thermo Scientific Chemicals Supplier No. H2669803

CAS: 3777-70-6 | C10H16O | 152.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3777-70-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00053134
InChI Key XBLCAKKYMZVLPU-UHFFFAOYSA-N
Synonym 2-n-hexylfuran, furan, 2-hexyl, 2-hexyl-furan, acmc-1clja
PubChem CID 77408
IUPAC Name 2-hexylfuran
SMILES CCCCCCC1=CC=CO1

Specifications

Quantity 1 g
UN Number UN1993
Formula Weight 152.24
Percent Purity 97%
Chemical Name or Material 2-n-Hexylfuran
Hazard Category H226-H302
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 223-235-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only