missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Heptylfuran, 97%
CAS: 3777-71-7 | C11H18O | 166.264 g/mol
$234.78 - $877.36
Chemical Identifiers
| CAS | 3777-71-7 |
|---|---|
| Molecular Formula | C11H18O |
| Molecular Weight (g/mol) | 166.264 |
| MDL Number | MFCD00051820 |
| InChI Key | BHTUFJXTYNLISA-UHFFFAOYSA-N |
| Synonym | 2-n-heptylfuran, furan, 2-heptyl, unii-7w55a39qxm, ccris 6901, fema no. 3401, 2-heptyl furan, furan, heptyl |
| PubChem CID | 19603 |
| IUPAC Name | 2-heptylfuran |
| SMILES | CCCCCCCC1=CC=CO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1060409
|
Thermo Scientific Chemicals
A1060409 |
10 g |
Each for $234.78
|
|
|||||
|
AAA1060418
|
Thermo Scientific Chemicals
A1060418 |
50 g |
Each for $877.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3777-71-7 | |
| 166.264 | |
| BHTUFJXTYNLISA-UHFFFAOYSA-N | |
| 19603 | |
| CCCCCCCC1=CC=CO1 |
| C11H18O | |
| MFCD00051820 | |
| 2-n-heptylfuran, furan, 2-heptyl, unii-7w55a39qxm, ccris 6901, fema no. 3401, 2-heptyl furan, furan, heptyl | |
| 2-heptylfuran |
Specifications
| 3777-71-7 | |
| 96°C to 98°C (30 mmHg) | |
| C11H18O | |
| MFCD00051820 | |
| 2-n-heptylfuran, furan, 2-heptyl, unii-7w55a39qxm, ccris 6901, fema no. 3401, 2-heptyl furan, furan, heptyl | |
| CCCCCCCC1=CC=CO1 | |
| 166.264 | |
| 166.27 | |
| 2-n-Heptylfuran |
| 0.863 | |
| 75°C (167°F) | |
| 1.4525 | |
| 10 g | |
| BHTUFJXTYNLISA-UHFFFAOYSA-N | |
| 2-heptylfuran | |
| 19603 | |
| 98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 223-236-8
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only