missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(N-Ethylanilino)ethanol 98.0+%, TCI America™
$67.11 - $246.05
Chemical Identifiers
| CAS | 92-50-2 |
|---|---|
| Molecular Formula | C10H15NO |
| Molecular Weight (g/mol) | 165.24 |
| MDL Number | MFCD00020575 |
| InChI Key | HYVGFUIWHXLVNV-UHFFFAOYSA-N |
| Synonym | 2-n-ethylanilino ethanol, 2-ethyl phenyl amino ethanol, ethanol, 2-ethylphenylamino, ethylphenylethanolamine, phenylethylethanolamine, n-ethyl-n-2-hydroxyethyl aniline, n-ethylanilinoethanol, hydroxyethylethylaniline, n-ethyl-n-hydroxyethylaniline, 2-ethylphenylamino ethanol |
| PubChem CID | 62338 |
| IUPAC Name | 2-[ethyl(phenyl)amino]ethan-1-ol |
| SMILES | CCN(CCO)C1=CC=CC=C1 |
Chemical Identifiers
| 92-50-2 | |
| 165.24 | |
| HYVGFUIWHXLVNV-UHFFFAOYSA-N | |
| 62338 | |
| CCN(CCO)C1=CC=CC=C1 |
| C10H15NO | |
| MFCD00020575 | |
| 2-n-ethylanilino ethanol, 2-ethyl phenyl amino ethanol, ethanol, 2-ethylphenylamino, ethylphenylethanolamine, phenylethylethanolamine, n-ethyl-n-2-hydroxyethyl aniline, n-ethylanilinoethanol, hydroxyethylethylaniline, n-ethyl-n-hydroxyethylaniline, 2-ethylphenylamino ethanol | |
| 2-[ethyl(phenyl)amino]ethan-1-ol |
Specifications
| 92-50-2 | |
| 268°C | |
| MFCD00020575 | |
| 2-n-ethylanilino ethanol, 2-ethyl phenyl amino ethanol, ethanol, 2-ethylphenylamino, ethylphenylethanolamine, phenylethylethanolamine, n-ethyl-n-2-hydroxyethyl aniline, n-ethylanilinoethanol, hydroxyethylethylaniline, n-ethyl-n-hydroxyethylaniline, 2-ethylphenylamino ethanol | |
| CCN(CCO)C1=CC=CC=C1 | |
| 165.24 | |
| 165.24 | |
| Crystalline Lumps |
| White-Yellow | |
| C10H15NO | |
| 25 g | |
| HYVGFUIWHXLVNV-UHFFFAOYSA-N | |
| 2-[ethyl(phenyl)amino]ethan-1-ol | |
| 62338 | |
| ≥98.0% (GC,T) | |
| 2-(N-Ethylanilino)ethanol |
Safety and Handling
EINECSNumber : (3)-0159
RTECSNumber : KL0878000
TSCA : Yes