Learn More
2-n-Butylthiophene, 98+%
CAS: 1455-20-5 | C8H12S | 140.244 g/mol
$104.86 - $372.48
Chemical Identifiers
| CAS | 1455-20-5 |
|---|---|
| Molecular Formula | C8H12S |
| Molecular Weight (g/mol) | 140.244 |
| MDL Number | MFCD00022534 |
| InChI Key | MNDZHERKKXUTOE-UHFFFAOYSA-N |
| Synonym | 2-n-butylthiophene, thiophene, 2-butyl, 2-n-butyl thiophene, ccris 6900, 2-butyl-thiophene, thiophene, butyl, acmc-20c3ak |
| PubChem CID | 73818 |
| IUPAC Name | 2-butylthiophene |
| SMILES | CCCCC1=CC=CS1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1274006
|
Thermo Scientific Chemicals
A1274006 |
5 g |
Each for $104.86
|
|
|||||
|
AAA1274014
|
Thermo Scientific Chemicals
A1274014 |
25 g |
Each for $372.48
|
|
|||||
Description
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.
Solubility
Soluble in acetone and DMSO.
Notes
Storage temperature: 2 - 4°Cand Keep container tightlyclosed in a dryand well-ventilated place. Incompatible to Strong oxidizing agents.
Chemical Identifiers
| 1455-20-5 | |
| 140.244 | |
| MNDZHERKKXUTOE-UHFFFAOYSA-N | |
| 73818 | |
| CCCCC1=CC=CS1 |
| C8H12S | |
| MFCD00022534 | |
| 2-n-butylthiophene, thiophene, 2-butyl, 2-n-butyl thiophene, ccris 6900, 2-butyl-thiophene, thiophene, butyl, acmc-20c3ak | |
| 2-butylthiophene |
Specifications
| 1455-20-5 | |
| 181°C to 182°C | |
| Recognizable | |
| 1.502 | |
| 5 g | |
| 107046 | |
| Soluble in acetone and DMSO. | |
| CCCCC1=CC=CS1 | |
| 140.244 | |
| 140.25 | |
| 2-n-Butylthiophene |
| 0.953 | |
| 56°C (133°F) | |
| C8H12S | |
| MFCD00022534 | |
| UN1993 | |
| 2-n-butylthiophene, thiophene, 2-butyl, 2-n-butyl thiophene, ccris 6900, 2-butyl-thiophene, thiophene, butyl, acmc-20c3ak | |
| MNDZHERKKXUTOE-UHFFFAOYSA-N | |
| 2-butylthiophene | |
| 73818 | |
| ≥98% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 215-935-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only