missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Butylthiophene, 98+%
Description
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.
Solubility
Soluble in acetone and DMSO.
Notes
Storage temperature: 2 - 4°Cand Keep container tightlyclosed in a dryand well-ventilated place. Incompatible to Strong oxidizing agents.
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.
Solubility
Soluble in acetone and DMSO.
Notes
Storage temperature: 2 - 4°Cand Keep container tightlyclosed in a dryand well-ventilated place. Incompatible to Strong oxidizing agents.
Specifications
Specifications
| Density | 0.953 |
| Boiling Point | 181°C to 182°C |
| Flash Point | 56°C (133°F) |
| Odor | Recognizable |
| Refractive Index | 1.502 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Beilstein | 107046 |
| Solubility Information | Soluble in acetone and DMSO. |
| Formula Weight | 140.25 |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H StatementH226Flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 215-935-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only