missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butylthiophene, 98+%

Catalog No. AAA1274006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274006 5 g
AAA1274014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1274006 Supplier Thermo Scientific Chemicals Supplier No. A1274006
Only null left

CAS: 1455-20-5 | C8H12S | 140.244 g/mol

2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

Solubility
Soluble in acetone and DMSO.

Notes
Storage temperature: 2 - 4°Cand Keep container tightlyclosed in a dryand well-ventilated place. Incompatible to Strong oxidizing agents.

Chemical Identifiers

CAS 1455-20-5
Molecular Formula C8H12S
Molecular Weight (g/mol) 140.244
MDL Number MFCD00022534
InChI Key MNDZHERKKXUTOE-UHFFFAOYSA-N
Synonym 2-n-butylthiophene, thiophene, 2-butyl, 2-n-butyl thiophene, ccris 6900, 2-butyl-thiophene, thiophene, butyl, acmc-20c3ak
PubChem CID 73818
IUPAC Name 2-butylthiophene
SMILES CCCCC1=CC=CS1

Specifications

Density 0.953
Boiling Point 181°C to 182°C
Flash Point 56°C (133°F)
Odor Recognizable
Refractive Index 1.502
Quantity 5 g
UN Number UN1993
Beilstein 107046
Solubility Information Soluble in acetone and DMSO.
Formula Weight 140.25
Percent Purity ≥98%
Chemical Name or Material 2-n-Butylthiophene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 215-935-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only