missing translation for 'onlineSavingsMsg'
Learn More

2-(Morpholinosulfonyl)benzylamine hydrochloride, 97%, Thermo Scientific™

Catalog No. CC69663DA
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC69663DA 1g Amber Glass Bottle
CC69663CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC69663DA Supplier Thermo Scientific Chemicals Supplier No. CC69663DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 918812-18-7
Molecular Formula C11H17ClN2O3S
Molecular Weight (g/mol) 292.778
MDL Number MFCD09284586
InChI Key RMMDNVBLYNZQFP-UHFFFAOYSA-N
Synonym 2-morpholinosulfonyl benzylamine hydrochloride, 1-2-morpholin-4-ylsulfonyl phenyl methanamine hydrochloride, 2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 1-2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride salt, 1-2-morpholine-4-sulfonyl phenyl methanamine-hydrogen chloride 1/1
PubChem CID 16320333
IUPAC Name (2-morpholin-4-ylsulfonylphenyl)methanamine;hydrochloride
SMILES C1COCCN1S(=O)(=O)C2=CC=CC=C2CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 292.79
Percent Purity 97%
Chemical Name or Material 2-(Morpholinosulfonyl)benzylamine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.