missing translation for 'onlineSavingsMsg'
Learn More

2-(morpholinosulfonyl)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC69601CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC69601CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

87223-34-5
271.287
YTQKWJNSSYODEV-UHFFFAOYSA-N
13140207
C1COCCN1S(=O)(=O)C2=CC=CC=C2C(=O)O
C11H13NO5S
MFCD09813541
2-morpholinosulfonyl benzoic acid, 2-morpholine-4-sulfonyl benzoic acid, 2-morpholin-4-ylsulfonyl benzoic acid, benzoic acid,2-4-morpholinylsulfonyl, 2-morpholin-4-ylsulphonyl benzoic acid
2-morpholin-4-ylsulfonylbenzoic acid

Specifications

2-(Morpholinosulfonyl)benzoic acid
96.5
97%
C11H13NO5S
MFCD09813541
YTQKWJNSSYODEV-UHFFFAOYSA-N
2-morpholin-4-ylsulfonylbenzoic acid
13140207
97%
87223-34-5
100.0
Amber Glass Bottle
250mg
2-morpholinosulfonyl benzoic acid, 2-morpholine-4-sulfonyl benzoic acid, 2-morpholin-4-ylsulfonyl benzoic acid, benzoic acid,2-4-morpholinylsulfonyl, 2-morpholin-4-ylsulphonyl benzoic acid
C1COCCN1S(=O)(=O)C2=CC=CC=C2C(=O)O
271.287
271.29
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents