missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinopyridine-4-boronic acid pinacol ester, 97%

Catalog No. AC445200050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445200050 5 g
AC445200010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445200050 Supplier Thermo Scientific Chemicals Supplier No. 445200050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 888721-86-6 | C15H23BN2O3 | 290.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 888721-86-6
Molecular Formula C15H23BN2O3
Molecular Weight (g/mol) 290.17
MDL Number MFCD11054039
InChI Key HZJIGVGLNZISJU-UHFFFAOYSA-N
Synonym 2-morpholinopyridine-4-boronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 2-morpholinopyridin-4-ylboronic acid pinacol ester, 2-morpholino pyridine-4-boronic acid, pinacol ester, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-morpholine
PubChem CID 21081661
IUPAC Name 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC(=NC=C1)N1CCOCC1

Specifications

Melting Point 85°C to 86°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 290.17
Percent Purity 97%
Chemical Name or Material 2-Morpholinopyridine-4-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only