missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC24309DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC24309DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

465514-33-4
193.25
MYGVYNRBQSAMIF-UHFFFAOYSA-N
2776562
OCC1=CC=CC=C1N1CCOCC1
C11H15NO2
MFCD03086181
2-morpholin-4-yl-phenyl methanol, 2-morpholinophenyl methanol, 2-morpholin-4-yl phenyl methanol, 2-morpholin-4-ylphenyl methanol, benzenemethanol,2-4-morpholinyl, 2-morpholinophenylmethanol, 2-morpholin-4-ylphenyl methan-1-ol
[2-(morpholin-4-yl)phenyl]methanol

Specifications

(2-Morpholinophenyl)methanol
96.5
97%
C11H15NO2
MFCD03086181
MYGVYNRBQSAMIF-UHFFFAOYSA-N
[2-(morpholin-4-yl)phenyl]methanol
2776562
97%
465514-33-4
100.0
Amber Glass Bottle
1g
2-morpholin-4-yl-phenyl methanol, 2-morpholinophenyl methanol, 2-morpholin-4-yl phenyl methanol, 2-morpholin-4-ylphenyl methanol, benzenemethanol,2-4-morpholinyl, 2-morpholinophenylmethanol, 2-morpholin-4-ylphenyl methan-1-ol
OCC1=CC=CC=C1N1CCOCC1
193.25
193.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions