missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinoaniline 98.0+%, TCI America™

Catalog No. p-7133362
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26715G 5 g
M267125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26715G Supplier TCI America Supplier No. M26715G
Only null left

Chemical Identifiers

CAS 5585-33-1
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00047408
InChI Key QKWLVAYDAHQMLG-UHFFFAOYSA-N
Synonym 2-morpholinoaniline, 2-morpholin-4-yl aniline, 2-morpholin-4-yl-phenylamine, 2-4-morpholino aniline, 4-2-aminophenyl morpholine, 2-4-morpholinyl aniline, 2-morpholin-4-ylphenylamine, enamine_000782, acmc-1ap2s
PubChem CID 735756
IUPAC Name 2-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=CC=C2N

Specifications

Melting Point 96°C
Color White-Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Morpholinoaniline