missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. p-3522461
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC57801CB 250mg Amber Glass Bottle
CC57801DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC57801CB Supplier Thermo Scientific Chemicals Supplier No. CC57801CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 865471-20-1
Molecular Formula C12H12F3NO3
Molecular Weight (g/mol) 275.227
MDL Number MFCD09025883
InChI Key PRECFTDWCNEEDB-UHFFFAOYSA-N
Synonym 2-morpholino-5-trifluoromethyl benzoic acid, 2-morpholin-4-yl-5-trifluoromethyl benzoic acid, benzoic acid,2-4-morpholinyl-5-trifluoromethyl
PubChem CID 24229572
IUPAC Name 2-morpholin-4-yl-5-(trifluoromethyl)benzoic acid
SMILES C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 275.22
Percent Purity 97%
Chemical Name or Material 2-Morpholino-5-(trifluoromethyl)benzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.