missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC57801DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC57801DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

865471-20-1
275.227
PRECFTDWCNEEDB-UHFFFAOYSA-N
24229572
C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)C(=O)O
C12H12F3NO3
MFCD09025883
2-morpholino-5-trifluoromethyl benzoic acid, 2-morpholin-4-yl-5-trifluoromethyl benzoic acid, benzoic acid,2-4-morpholinyl-5-trifluoromethyl
2-morpholin-4-yl-5-(trifluoromethyl)benzoic acid

Specifications

2-Morpholino-5-(trifluoromethyl)benzoic acid
96.5
97%
C12H12F3NO3
MFCD09025883
PRECFTDWCNEEDB-UHFFFAOYSA-N
2-morpholin-4-yl-5-(trifluoromethyl)benzoic acid
24229572
97%
865471-20-1
100.0
Amber Glass Bottle
1g
2-morpholino-5-trifluoromethyl benzoic acid, 2-morpholin-4-yl-5-trifluoromethyl benzoic acid, benzoic acid,2-4-morpholinyl-5-trifluoromethyl
C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)C(=O)O
275.227
275.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents