missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholino-3-pyridinyl)methanol, 97%, Thermo Scientific™

$239.62 - $239.62

Chemical Identifiers

CAS 423768-55-2
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.23
MDL Number MFCD03086196
InChI Key WESHZTWFDUKWGL-UHFFFAOYSA-N
PubChem CID 2776571
IUPAC Name (2-morpholin-4-ylpyridin-3-yl)methanol
SMILES OCC1=C(N=CC=C1)N1CCOCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC24609DA
SDS View Documents
Thermo Scientific Chemicals
CC24609DA
1 g Amber Glass Bottle
Each for $239.62
Only null left
 
Description

Description

Specifications

Chemical Identifiers

423768-55-2
194.23
WESHZTWFDUKWGL-UHFFFAOYSA-N
(2-morpholin-4-ylpyridin-3-yl)methanol
C10H14N2O2
MFCD03086196
2776571
OCC1=C(N=CC=C1)N1CCOCC1

Specifications

423768-55-2
100.0
Amber Glass Bottle
MFCD03086196
2-morpholino-3-pyridinyl methanol, 2-morpholin-4-yl pyridin-3-yl methanol, 3-pyridinemethanol,2-4-morpholinyl, 2-morpholinopyridin-3-yl methanol, 3-pyridinemethanol, 2-4-morpholinyl, 2-morpholin-4-ylpyridin-3-yl methanol, 2-morpholin-4-yl-3-pyridyl methan-1-ol
OCC1=C(N=CC=C1)N1CCOCC1
194.23
194.23
(2-Morpholino-3-pyridinyl)methanol
96.5
97%
C10H14N2O2
1 g
WESHZTWFDUKWGL-UHFFFAOYSA-N
(2-morpholin-4-ylpyridin-3-yl)methanol
2776571
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'