missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholino-3-pyridinyl)methanol, 97%, Thermo Scientific™

Catalog No. CC24609DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24609DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC24609DA Supplier Thermo Scientific Chemicals Supplier No. CC24609DA

Chemical Identifiers

CAS 423768-55-2
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.23
MDL Number MFCD03086196
InChI Key WESHZTWFDUKWGL-UHFFFAOYSA-N
PubChem CID 2776571
IUPAC Name (2-morpholin-4-ylpyridin-3-yl)methanol
SMILES OCC1=C(N=CC=C1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-morpholino-3-pyridinyl methanol, 2-morpholin-4-yl pyridin-3-yl methanol, 3-pyridinemethanol,2-4-morpholinyl, 2-morpholinopyridin-3-yl methanol, 3-pyridinemethanol, 2-4-morpholinyl, 2-morpholin-4-ylpyridin-3-yl methanol, 2-morpholin-4-yl-3-pyridyl methan-1-ol
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material (2-Morpholino-3-pyridinyl)methanol
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.