missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-3-pyridinamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 51627-47-5
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD03001243
InChI Key DHYQHRDXKCKJAN-UHFFFAOYSA-N
Synonym 2-morpholinopyridin-3-amine, 2-morpholino-3-pyridinamine, 2-morpholin-4-yl pyridin-3-amine, 2-morpholin-4-yl-3-pyridylamine, 2-morpholin-4-yl-pyridin-3-ylamine, 3-amino-2-morpholinopyridine, 3-pyridinamine,2-4-morpholinyl
PubChem CID 2776573
IUPAC Name 2-morpholin-4-ylpyridin-3-amine
SMILES C1COCCN1C2=C(C=CC=N2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC24614DA
SDS View Documents
Thermo Scientific Chemicals
CC24614DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC24614EA
SDS View Documents
Thermo Scientific Chemicals
CC24614EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

51627-47-5
179.223
DHYQHRDXKCKJAN-UHFFFAOYSA-N
2776573
C1COCCN1C2=C(C=CC=N2)N
C9H13N3O
MFCD03001243
2-morpholinopyridin-3-amine, 2-morpholino-3-pyridinamine, 2-morpholin-4-yl pyridin-3-amine, 2-morpholin-4-yl-3-pyridylamine, 2-morpholin-4-yl-pyridin-3-ylamine, 3-amino-2-morpholinopyridine, 3-pyridinamine,2-4-morpholinyl
2-morpholin-4-ylpyridin-3-amine

Specifications

51627-47-5
100.0
Amber Glass Bottle
MFCD03001243
2-morpholinopyridin-3-amine, 2-morpholino-3-pyridinamine, 2-morpholin-4-yl pyridin-3-amine, 2-morpholin-4-yl-3-pyridylamine, 2-morpholin-4-yl-pyridin-3-ylamine, 3-amino-2-morpholinopyridine, 3-pyridinamine,2-4-morpholinyl
C1COCCN1C2=C(C=CC=N2)N
179.223
179.22
2-Morpholino-3-pyridinamine
96.5
97%
C9H13N3O
1g
DHYQHRDXKCKJAN-UHFFFAOYSA-N
2-morpholin-4-ylpyridin-3-amine
2776573
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents