missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiophene 97.0+%, TCI America™

Catalog No. M063525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M063525ML 25 mL
M0635500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M063525ML Supplier TCI America Supplier No. M063525ML

Chemical Identifiers

CAS 554-14-3
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005451
InChI Key XQQBUAPQHNYYRS-UHFFFAOYSA-N
Synonym thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
PubChem CID 11126
IUPAC Name 2-methylthiophene
SMILES CC1=CC=CS1

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 113°C
Quantity 25 mL
UN Number 1993
Formula Weight 98.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylthiophene
RTECSNumber XM9625000
TSCA Yes