missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylthiophene, 98%
Description
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;
Solubility
Slightly soluble in water. Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases, strong oxidizing agents.
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;
Solubility
Slightly soluble in water. Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases, strong oxidizing agents.
Specifications
Specifications
| Melting Point | -63°C |
| Density | 1.017 |
| Boiling Point | 112°C to 114°C |
| Flash Point | 7°C (44°F) |
| Odor | Aromatic |
| Refractive Index | 1.52 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Beilstein | 103734 |
| Solubility Information | Slightly soluble in water. Soluble in alcohol. |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H225-H315-H319-H332-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H225-H332 Highly flammable liquid and vapour. Harmful if inhaled. |
| missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 209-063-0 |
| RTECSNumber | XM9625000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only