missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)nicotinic acid, 98+%

Catalog No. AAL0992004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0992004 2 g
AAL0992009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0992004 Supplier Thermo Scientific Chemicals Supplier No. L0992004

CAS: 74470-23-8 | C7H7NO2S | 169.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74470-23-8
Molecular Formula C7H7NO2S
Molecular Weight (g/mol) 169.198
MDL Number MFCD00010101
InChI Key COPSJQVPEUUOKY-UHFFFAOYSA-N
Synonym 2-methylthio nicotinic acid, 2-methylsulfanyl nicotinic acid, 2-methylsulfanyl pyridine-3-carboxylic acid, 2-methylthio pyridine-3-carboxylic acid, 3-pyridinecarboxylic acid, 2-methylthio, 2-methylthio-3-pyridinecarboxylic acid, 2-methylmercapto nicotinic acid, 2-methylthiopyridine-3-carboxylic acid, acmc-20aomm, maybridge1_001445
PubChem CID 262400
IUPAC Name 2-methylsulfanylpyridine-3-carboxylic acid
SMILES CSC1=C(C=CC=N1)C(=O)O

Specifications

Melting Point 215°C to 218°C
Quantity 2 g
Beilstein 4391503
Formula Weight 169.2
Percent Purity ≥98%
Chemical Name or Material 2-(Methylthio)nicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only