missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylthio)benzoxazole 98.0+%, TCI America™
$109.74 - $429.31
Chemical Identifiers
| CAS | 13673-62-6 |
|---|---|
| Molecular Formula | C8H7NOS |
| Molecular Weight (g/mol) | 165.21 |
| MDL Number | MFCD00160027 |
| InChI Key | CBXAWZGFEDZKFR-UHFFFAOYSA-N |
| PubChem CID | 4178773 |
| IUPAC Name | 2-methylsulfanyl-1,3-benzoxazole |
| SMILES | CSC1=NC2=CC=CC=C2O1 |
Chemical Identifiers
| 13673-62-6 | |
| 165.21 | |
| CBXAWZGFEDZKFR-UHFFFAOYSA-N | |
| 2-methylsulfanyl-1,3-benzoxazole |
| C8H7NOS | |
| MFCD00160027 | |
| 4178773 | |
| CSC1=NC2=CC=CC=C2O1 |
Specifications
| 13673-62-6 | |
| 122°C | |
| MFCD00160027 | |
| CBXAWZGFEDZKFR-UHFFFAOYSA-N | |
| 2-methylsulfanyl-1,3-benzoxazole | |
| 4178773 | |
| ≥98.0% (GC) | |
| 2-(Methylthio)benzoxazole |
| Yellow | |
| C8H7NOS | |
| 1 g | |
| CSC1=NC2=CC=CC=C2O1 | |
| 165.21 | |
| 165.21 | |
| Liquid |
Safety and Handling
TSCA : No