missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylthio)acetamide 98.0+%, TCI America™
$34.66 - $94.33
Chemical Identifiers
| CAS | 22551-24-2 |
|---|---|
| Molecular Formula | C3H7NOS |
| Molecular Weight (g/mol) | 105.155 |
| MDL Number | MFCD00041321 |
| InChI Key | OBENGAMZEGRSIC-UHFFFAOYSA-N |
| PubChem CID | 547873 |
| IUPAC Name | 2-methylsulfanylacetamide |
| SMILES | CSCC(=O)N |
Chemical Identifiers
| 22551-24-2 | |
| 105.155 | |
| OBENGAMZEGRSIC-UHFFFAOYSA-N | |
| 2-methylsulfanylacetamide |
| C3H7NOS | |
| MFCD00041321 | |
| 547873 | |
| CSCC(=O)N |
Specifications
| 22551-24-2 | |
| White | |
| MFCD00041321 | |
| OBENGAMZEGRSIC-UHFFFAOYSA-N | |
| 2-methylsulfanylacetamide | |
| 547873 | |
| ≥98.0% (GC) | |
| 2-(Methylthio)acetamide |
| 104°C | |
| C3H7NOS | |
| 1 g | |
| CSCC(=O)N | |
| 105.155 | |
| 105.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No