missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)acetamide 98.0+%, TCI America™

Catalog No. M25791G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25791G 1 g
M25795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25791G Supplier TCI America Supplier No. M25791G

Chemical Identifiers

CAS 22551-24-2
Molecular Formula C3H7NOS
Molecular Weight (g/mol) 105.155
MDL Number MFCD00041321
InChI Key OBENGAMZEGRSIC-UHFFFAOYSA-N
PubChem CID 547873
IUPAC Name 2-methylsulfanylacetamide
SMILES CSCC(=O)N

Specifications

Melting Point 104°C
Color White
Quantity 1 g
Formula Weight 105.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)acetamide