missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one 98.0+%, TCI America™

Catalog No. M28515G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28515G 5 g
M28511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28515G Supplier TCI America Supplier No. M28515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16097-62-4
Molecular Formula C6H5F3N2OS
Molecular Weight (g/mol) 210.174
MDL Number MFCD00153191
InChI Key BRLPDRZKOMZVOY-UHFFFAOYSA-N
Synonym 2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
PubChem CID 248062
IUPAC Name 2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES CSC1=NC(=O)C=C(N1)C(F)(F)F

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 210.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one