missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylthio-4-pyrimidinol 98.0+%, TCI America™
$33.56 - $91.33
Chemical Identifiers
| CAS | 5751-20-2 |
|---|---|
| Molecular Formula | C5H6N2OS |
| Molecular Weight (g/mol) | 142.176 |
| MDL Number | MFCD00047373 |
| InChI Key | UYHSQVMHSFXUOA-UHFFFAOYSA-N |
| Synonym | 2-methylthio pyrimidin-4-ol, 2-methylthiopyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio pyrimidin-4 3h-one, 2-methylthio-4-pyrimidone, 2-methylthiouracil, 2-methylsulfanyl pyrimidin-4-ol, 2-methylthio-4-hydroxypyrimidine, 4 1h-pyrimidinone, 2-methylthio |
| PubChem CID | 79823 |
| IUPAC Name | 2-methylsulfanyl-1H-pyrimidin-6-one |
| SMILES | CSC1=NC=CC(=O)N1 |
Chemical Identifiers
| 5751-20-2 | |
| 142.176 | |
| UYHSQVMHSFXUOA-UHFFFAOYSA-N | |
| 79823 | |
| CSC1=NC=CC(=O)N1 |
| C5H6N2OS | |
| MFCD00047373 | |
| 2-methylthio pyrimidin-4-ol, 2-methylthiopyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio pyrimidin-4 3h-one, 2-methylthio-4-pyrimidone, 2-methylthiouracil, 2-methylsulfanyl pyrimidin-4-ol, 2-methylthio-4-hydroxypyrimidine, 4 1h-pyrimidinone, 2-methylthio | |
| 2-methylsulfanyl-1H-pyrimidin-6-one |
Specifications
| 5751-20-2 | |
| White-Yellow | |
| MFCD00047373 | |
| 2-methylthio pyrimidin-4-ol, 2-methylthiopyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio pyrimidin-4 3h-one, 2-methylthio-4-pyrimidone, 2-methylthiouracil, 2-methylsulfanyl pyrimidin-4-ol, 2-methylthio-4-hydroxypyrimidine, 4 1h-pyrimidinone, 2-methylthio | |
| CSC1=NC=CC(=O)N1 | |
| 142.176 | |
| 142.18 | |
| Crystalline Powder |
| 202°C | |
| C5H6N2OS | |
| 1 g | |
| UYHSQVMHSFXUOA-UHFFFAOYSA-N | |
| 2-methylsulfanyl-1H-pyrimidin-6-one | |
| 79823 | |
| ≥98.0% (HPLC,T) | |
| 2-Methylthio-4-pyrimidinol |
Safety and Handling
TSCA : No