missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthio-4-pyrimidinol 98.0+%, TCI America™

Catalog No. M23555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23555G 5 g
M23551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23555G Supplier TCI America Supplier No. M23555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5751-20-2
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.176
MDL Number MFCD00047373
InChI Key UYHSQVMHSFXUOA-UHFFFAOYSA-N
Synonym 2-methylthio pyrimidin-4-ol, 2-methylthiopyrimidin-4-ol, 2-methylthio-4-pyrimidinol, 2-methylthio pyrimidin-4 3h-one, 2-methylthio-4-pyrimidone, 2-methylthiouracil, 2-methylsulfanyl pyrimidin-4-ol, 2-methylthio-4-hydroxypyrimidine, 4 1h-pyrimidinone, 2-methylthio
PubChem CID 79823
IUPAC Name 2-methylsulfanyl-1H-pyrimidin-6-one
SMILES CSC1=NC=CC(=O)N1

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 5 g
Formula Weight 142.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylthio-4-pyrimidinol