missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiazoline 98.0+%, TCI America™

Supplier:  TCI America M0285100ML

 View more versions of this product

Catalog No. M0285100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methylthiazoline
Yellow
C4H7NS
100 mL
2-methyl-2-thiazoline, 2-methylthiazoline, thiazole, 4,5-dihydro-2-methyl, 2-methyl-4,5-dihydrothiazole, 4,5-dihydro-2-methylthiazole, methyl-2-thiazoline, 2-thiazoline, 2-methyl, unii-8z6ua8v7wn, methyl-2 delta-2 thiazoline, methyl-2 delta-2 thiazoline french
CC1=NCCS1
101.17
101.17
Liquid
2346-00-1
145°C
MFCD00005314
1993
JUIQOABNSLTJSW-UHFFFAOYSA-N
2-methyl-4,5-dihydro-1,3-thiazole
16867
≥98.0% (GC)

Chemical Identifiers

2346-00-1
101.17
JUIQOABNSLTJSW-UHFFFAOYSA-N
16867
CC1=NCCS1
C4H7NS
MFCD00005314
2-methyl-2-thiazoline, 2-methylthiazoline, thiazole, 4,5-dihydro-2-methyl, 2-methyl-4,5-dihydrothiazole, 4,5-dihydro-2-methylthiazole, methyl-2-thiazoline, 2-thiazoline, 2-methyl, unii-8z6ua8v7wn, methyl-2 delta-2 thiazoline, methyl-2 delta-2 thiazoline french
2-methyl-4,5-dihydro-1,3-thiazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0215

RTECSNumber : XJ4261470

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions