missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiazoline 98.0+%, TCI America™

Catalog No. M028525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M028525ML 25 mL
M0285100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M028525ML Supplier TCI America Supplier No. M028525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2346-00-1
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.17
MDL Number MFCD00005314
InChI Key JUIQOABNSLTJSW-UHFFFAOYSA-N
Synonym 2-methyl-2-thiazoline, 2-methylthiazoline, thiazole, 4,5-dihydro-2-methyl, 2-methyl-4,5-dihydrothiazole, 4,5-dihydro-2-methylthiazole, methyl-2-thiazoline, 2-thiazoline, 2-methyl, unii-8z6ua8v7wn, methyl-2 delta-2 thiazoline, methyl-2 delta-2 thiazoline french
PubChem CID 16867
IUPAC Name 2-methyl-4,5-dihydro-1,3-thiazole
SMILES CC1=NCCS1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 1993
Formula Weight 101.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylthiazoline
EINECSNumber (5)-0215
RTECSNumber XJ4261470
TSCA Yes
Recommended Storage Refrigerator