missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylthiazole-5-carboxylic Acid 98.0+%, TCI America™
$186.74 - $750.82
Chemical Identifiers
| CAS | 40004-69-1 |
|---|---|
| Molecular Formula | C5H5NO2S |
| Molecular Weight (g/mol) | 143.16 |
| MDL Number | MFCD01760300 |
| InChI Key | QCXCIYPOMMIBHO-UHFFFAOYSA-N |
| PubChem CID | 98845 |
| IUPAC Name | 2-methyl-1,3-thiazole-5-carboxylic acid |
| SMILES | CC1=NC=C(S1)C(=O)O |
Chemical Identifiers
| 40004-69-1 | |
| 143.16 | |
| QCXCIYPOMMIBHO-UHFFFAOYSA-N | |
| 2-methyl-1,3-thiazole-5-carboxylic acid |
| C5H5NO2S | |
| MFCD01760300 | |
| 98845 | |
| CC1=NC=C(S1)C(=O)O |
Specifications
| 40004-69-1 | |
| C5H5NO2S | |
| 1 g | |
| CC1=NC=C(S1)C(=O)O | |
| 143.16 | |
| 143.16 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01760300 | |
| QCXCIYPOMMIBHO-UHFFFAOYSA-N | |
| 2-methyl-1,3-thiazole-5-carboxylic acid | |
| 98845 | |
| ≥98.0% (GC,T) | |
| 2-Methylthiazole-5-carboxylic Acid |
Safety and Handling
RTECSNumber : XJ3678500
TSCA : No