missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, ReagentPlus™, ≥99.5%, Honeywell Burdick & Jackson™

Catalog No. p-7120073
Change view
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044898 2.5 L Glass Bottle
60044900 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044898 Supplier Honeywell Inc Supplier No. 1558102.5L
Only null left

Reagent Grade, contains 150-400 ppm BHT as stabilizer,≥99.5%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00005367
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Melting Point <-20°C
Color Colorless
Density 0.86g/mL at 25°C
Boiling Point 78°C to 80°C
Quantity 2.5 L
Flash Point −10°C (14°F)
Packaging Glass Bottle
Refractive Index n20/D 1.406(lit.)
UN Number UN2536
Beilstein 102448
Formula Weight 86.13g/mol
Percent Purity ≥99.5%
Grade ReagentPlus
Chemical Name or Material 2-Methyltetrahydrofuran
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P305 + P351 + P338 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H302-H315-H318
DOTInformation 3/PGII
EINECSNumber 202-507-4
RTECSNumber LU2800000