missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized

Catalog No. AC392380100
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392380100 10 L Metal Drum
AC392382500 250 mL Glass bottle
AC392380010 1 L Glass bottle
AC392380025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC392380100 Supplier Thermo Scientific Chemicals Supplier No. 392380100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Quantity: 2.5L, Packaging: Glass bottle, Boiling Point: 78.0 deg.C to 80.0 deg.C, CAS: 96-47-9, Colorless, Grade: For Grignard reaction, Melting Point: -136.0 deg.C, Molecular Formula: C5H10O | CAS: 96-47-9 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Melting Point -136.0°C
Color Colorless
Density 0.8600g/mL
Quantity 10 L
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.4046 to 1.4066
Specific Gravity 0.86
Viscosity 4 mPa.s (25°C)
Merck Index 15,6198
Boiling Point 78.0°C to 80.0°C
Solubility Information Solubility in water: 150g/l (25°C)
Formula Weight 86.13
Percent Purity ≥99.0%
Chemical Name or Material 2-Methyltetrahydrofuran, Stabilized; For Grignard reaction
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
EINECSNumber 202-507-4

RUO – Research Use Only