missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, 99%, stab. with ca 150-400ppm BHT

CAS: 96-47-9 | C5H10O | 86.134 g/mol

Supplier:  Thermo Scientific Chemicals L07356AP

 View more versions of this product

Catalog No. AAL07356AP


Only null left
Explore available promotions

Description

Description

2-Methyltetrahydrofuran acts as a solvent in organic synthesis. It is considered as a replacement for terahydrofuran due to its higher reaction temperature and easy separation after reaction. It is also useful in the electrolyte formulation for secondary lithium electrodes and as a component in alternative fuels. Further, it is used as a solvent for spectroscopic studies at -1960C. It also acts as a solvent for Grignard reagent in organometallic reactions. In addition to this, it plays an important role as a motor fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyltetrahydrofuran acts as a solvent in organic synthesis. It is considered as a replacement for terahydrofuran due to its higher reaction temperature and easy separation after reaction. It is also useful in the electrolyte formulation for secondary lithium electrodes and as a component in alternative fuels. Further, it is used as a solvent for spectroscopic studies at -1960C. It also acts as a solvent for Grignard reagent in organometallic reactions. In addition to this, it plays an important role as a motor fuel.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with oxidizing agents, strong acids and strong bases.
Specifications

Specifications

2-Methyltetrahydrofuran
stabilized with ≈150 to 400ppm BHT
0.856
500 mL
C5H10O
UN2536
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyloxolane
7301
99%
-136°C
96-47-9
80°C to 82°C
1.406
MFCD00005367
102448
Miscible with water and most organic solvents.
CC1CCCO1
86.134
86.13

Chemical Identifiers

96-47-9
86.134
JWUJQDFVADABEY-UHFFFAOYSA-N
7301
CC1CCCO1
C5H10O
MFCD00005367
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
2-methyloxolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302-H315-H318-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLTETRAHYDROFURAN

EINECSNumber : 202-507-4

RTECSNumber : LU2800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only