missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -136.0°C |
| Color | Colorless |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Density | 0.8600g/mL |
| Boiling Point | 78.0°C to 80.0°C |
| Quantity | 100 mL |
| Infrared Spectrum | Authentic |
| Packaging | AcroSeal™ Glass Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Harmful if swallowed. Causes skin irritation. Causes serious eye damage. May form explosive peroxides. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Wear protective g |
| EINECSNumber | 202-507-4 |
RUO – Research Use Only