missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, 99+%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™

CAS: 96-47-9 | C5H10O | 86.13 g/mol

Supplier:  Thermo Scientific Chemicals 396630010

 View more versions of this product

Catalog No. AC396630010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyltetrahydrofuran
Over Molecular Sieves, Stabilized
Colorless
100.0
78.0°C to 80.0°C
Authentic
Liquid
99% min. (GC)
C5H10O
4 mPa.s (25°C)
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyloxolane
7301
99+%
-136.0°C
96-47-9
99.0
0.8600g/mL
1 L
AcroSeal™ Glass Bottle
1.4046 to 1.4066
0.86
MFCD00005367
15, 6198
Solubility in water: 150g/l (25°C)
CC1CCCO1
86.13
86.13
Extra Dry over Molecular Sieve

Chemical Identifiers

96-47-9
86.13
JWUJQDFVADABEY-UHFFFAOYSA-N
7301
CC1CCCO1
C5H10O
MFCD00005367
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
2-methyloxolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g

GHS Signal Word: Danger

EINECSNumber : 202-507-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only