missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran-3-one, 98+%

Catalog No. AAB2089014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2089014 25 g
AAB2089006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2089014 Supplier Thermo Scientific Chemicals Supplier No. B2089014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3188-00-9 | C5H8O2 | 100.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3188-00-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00010423
InChI Key FCWYQRVIQDNGBI-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran-3-one, 2-methyl tetrahydro-3-furanone, 2-methyltetrahydro-3-furanone, 3 2h-furanone, dihydro-2-methyl, dihydro-2-methyl-3 2h-furanone, coffee furanone, dihydro-2-methyl-3-furanone, tetrahydro-2-methylfuran-3-one, 2-methyl-3-oxotetrahydrofuran, 2-methyl-3-ketotetrahydrofuran
PubChem CID 18522
IUPAC Name 2-methyloxolan-3-one
SMILES CC1C(=O)CCO1

Specifications

Density 1.034
Quantity 25 g
Odor Characteristic
Refractive Index 1.429
UN Number UN1224
Beilstein 1341334
Boiling Point 138°C to 139°C
Formula Weight 100.12
Percent Purity ≥98%
Chemical Name or Material 2-Methyltetrahydrofuran-3-one
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 221-685-4
RTECSNumber LU3579000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only