missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)acetophenone 98.0+%, TCI America™

Supplier:  TCI America M07125G

Catalog No. M07125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(Methylsulfonyl)acetophenone
108°C
C9H10O3S
5 g
CAUSPZIZBLGLKW-UHFFFAOYSA-N
2-methylsulfonyl-1-phenylethanone
19457
≥98.0% (GC)
3708-04-1
White-Yellow
MFCD00025078
Benzoylmethyl Methyl Sulfone, Methyl Phenacyl Sulfone
CS(=O)(=O)CC(=O)C1=CC=CC=C1
198.24
198.24
Crystalline Powder

Chemical Identifiers

3708-04-1
198.24
CAUSPZIZBLGLKW-UHFFFAOYSA-N
19457
CS(=O)(=O)CC(=O)C1=CC=CC=C1
C9H10O3S
MFCD00025078
Benzoylmethyl Methyl Sulfone, Methyl Phenacyl Sulfone
2-methylsulfonyl-1-phenylethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AM9486150

TSCA : No

SDS
Documents

Documents

Promotions

Promotions