Learn More
2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine, 96%, Thermo Scientific™
$673.60 - $673.60
Chemical Identifiers
| CAS | 122476-85-1 |
|---|---|
| Molecular Formula | C17H32N2O2SSn |
| Molecular Weight (g/mol) | 447.225 |
| MDL Number | MFCD07787379 |
| InChI Key | RRHTVQOGUAOHDR-UHFFFAOYSA-N |
| Synonym | 2-methylsulfonyl-5-tributylstannyl pyrimidine, 2-methanesulfonyl-5-tributylstannyl pyrimidine, 2-methylsulphonyl-5-tributylstannyl pyrimidine, tributyl-2-methylsulfonylpyrimidin-5-yl stannane, 2-methylsulfonyl-5-tri-n-butylstannyl pyrimidine |
| PubChem CID | 2763272 |
| IUPAC Name | tributyl-(2-methylsulfonylpyrimidin-5-yl)stannane |
| SMILES | CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(=O)(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5157403
|
Thermo Scientific Chemicals
H5157403 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 122476-85-1 | |
| 447.225 | |
| RRHTVQOGUAOHDR-UHFFFAOYSA-N | |
| 2763272 | |
| CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(=O)(=O)C |
| C17H32N2O2SSn | |
| MFCD07787379 | |
| 2-methylsulfonyl-5-tributylstannyl pyrimidine, 2-methanesulfonyl-5-tributylstannyl pyrimidine, 2-methylsulphonyl-5-tributylstannyl pyrimidine, tributyl-2-methylsulfonylpyrimidin-5-yl stannane, 2-methylsulfonyl-5-tri-n-butylstannyl pyrimidine | |
| tributyl-(2-methylsulfonylpyrimidin-5-yl)stannane |
Specifications
| 122476-85-1 | |
| MFCD07787379 | |
| UN2788 | |
| RRHTVQOGUAOHDR-UHFFFAOYSA-N | |
| tributyl-(2-methylsulfonylpyrimidin-5-yl)stannane | |
| 2763272 | |
| 96% |
| C17H32N2O2SSn | |
| 1 g | |
| 2-methylsulfonyl-5-tributylstannyl pyrimidine, 2-methanesulfonyl-5-tributylstannyl pyrimidine, 2-methylsulphonyl-5-tributylstannyl pyrimidine, tributyl-2-methylsulfonylpyrimidin-5-yl stannane, 2-methylsulfonyl-5-tri-n-butylstannyl pyrimidine | |
| CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(=O)(=O)C | |
| 447.225 | |
| 447.22 | |
| 2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Keep cold