missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine, 96%, Thermo Scientific™

Catalog No. AAH5157403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5157403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH5157403 Supplier Thermo Scientific Chemicals Supplier No. H5157403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 122476-85-1
Molecular Formula C17H32N2O2SSn
Molecular Weight (g/mol) 447.225
MDL Number MFCD07787379
InChI Key RRHTVQOGUAOHDR-UHFFFAOYSA-N
Synonym 2-methylsulfonyl-5-tributylstannyl pyrimidine, 2-methanesulfonyl-5-tributylstannyl pyrimidine, 2-methylsulphonyl-5-tributylstannyl pyrimidine, tributyl-2-methylsulfonylpyrimidin-5-yl stannane, 2-methylsulfonyl-5-tri-n-butylstannyl pyrimidine
PubChem CID 2763272
IUPAC Name tributyl-(2-methylsulfonylpyrimidin-5-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(=O)(=O)C

Specifications

Quantity 1 g
UN Number UN2788
Formula Weight 447.22
Percent Purity 96%
Chemical Name or Material 2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Keep cold