missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsulfanylpyrimidine-4-carboxylic acid, 97%

Catalog No. p-4228828
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432370010 1 g Glass bottle
AC432370050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432370010 Supplier Thermo Scientific Chemicals Supplier No. 432370010

CAS: 1126-44-9 | C6H6N2O2S | 170.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1126-44-9
Molecular Formula C6H6N2O2S
Molecular Weight (g/mol) 170.19
MDL Number MFCD09834808
InChI Key IAGNLKODEFUQDV-UHFFFAOYSA-N
Synonym 2-methylthio pyrimidine-4-carboxylic acid, 2-methylsulfanyl-pyrimidine-4-carboxylic acid, 2-methylthiopyrimidine-4-carboxylic acid, 2-thiomethylpyrimidine-4-carboxylic acid, 2-methylsulfanyl pyrimidine-4-carboxylic acid, 2-methylsulfanyl-pyrimidine-4-carboxylicacid, 2-methylthio-4-pyrimidinecarboxylic acid, 2-thioformylpyrimidine-4-carboxylic acid, 2-methylthio-4-pyrimidinecarboxylicacid, 2-mthylsulfanyl-pyrimidine-4-carboxylic acid
PubChem CID 295775
IUPAC Name 2-methylsulfanylpyrimidine-4-carboxylic acid
SMILES CSC1=NC=CC(=N1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 208°C to 209°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 170.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylsulfanylpyrimidine-4-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only