missing translation for 'onlineSavingsMsg'
Learn More

2-Methylstilbene 98.0+%, TCI America™

Catalog No. m14915g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14915G Supplier TCI America Supplier No. M14915G
Only null left

Chemical Identifiers

CAS 74685-42-0
Molecular Formula C15H14
Molecular Weight (g/mol) 194.28
MDL Number MFCD06797130
InChI Key PEBXLTUWFWEWGV-UHFFFAOYSA-N
PubChem CID 5375376
IUPAC Name 1-methyl-2-(2-phenylethenyl)benzene
SMILES CC1=CC=CC=C1C=CC1=CC=CC=C1

Specifications

Melting Point 33°C
Color White
Quantity 5 g
Formula Weight 194.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylstilbene
TSCA No
Recommended Storage Refrigerator