missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoline-6-carboxylic acid, 97%

CAS: 635-80-3 | C11H9NO2 | 187.198 g/mol

Supplier:  Thermo Scientific Chemicals H6379006

 View more versions of this product

Catalog No. AAH6379006

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylquinoline-6-carboxylic acid
C11H9NO2
5 g
IZONZQFTYGVOOO-UHFFFAOYSA-N
2-methylquinoline-6-carboxylic acid
604483
97%
635-80-3
MFCD00014566
2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid
CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O
187.198
187.19
Solid

Chemical Identifiers

635-80-3
187.198
IZONZQFTYGVOOO-UHFFFAOYSA-N
604483
CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O
C11H9NO2
MFCD00014566
2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid
2-methylquinoline-6-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only