missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoline-6-carboxylic acid, 97%

Catalog No. AAH6379003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6379003 1 g
AAH6379006 5 g
AAH6379014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6379003 Supplier Thermo Scientific Chemicals Supplier No. H6379003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 635-80-3 | C11H9NO2 | 187.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 635-80-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD00014566
InChI Key IZONZQFTYGVOOO-UHFFFAOYSA-N
Synonym 2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid
PubChem CID 604483
IUPAC Name 2-methylquinoline-6-carboxylic acid
SMILES CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O

Specifications

Quantity 1 g
Formula Weight 187.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Methylquinoline-6-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only