missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrimidine, 98%

Catalog No. AC459860010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459860010 1 g
AC459860050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459860010 Supplier Thermo Scientific Chemicals Supplier No. 459860010

CAS: 5053-43-0 | C5H6N2 | 94.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5053-43-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.11
InChI Key LNJMHEJAYSYZKK-UHFFFAOYSA-N
Synonym pyrimidine, 2-methyl, 2-methyl pyrimidine, pubchem21974, 2-methyl-1,3-diazine, 2-methylpyrimidine, pyrimidine, 2-methyl-6ci,7ci,8ci,9ci
PubChem CID 78748
IUPAC Name 2-methylpyrimidine
SMILES CC1=NC=CC=N1

Specifications

Melting Point -6.0°C
Density 1.0132g/mL
Boiling Point 130.0°C to 132.0°C
Flash Point 37.8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.0132
Formula Weight 94.11
Percent Purity 98%
Chemical Name or Material 2-Methylpyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

RUO – Research Use Only