missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpyridine 98.0+%, TCI America™
Supplier: TCI America P0415500ML
Specifications
| 2-Methylpyridine | |
| -70°C | |
| 128°C | |
| MFCD00006332 | |
| 2313 | |
| BSKHPKMHTQYZBB-UHFFFAOYSA-N | |
| 2-methylpyridine | |
| 7975 | |
| 93.13 | |
| Liquid |
| 109-06-8 | |
| Yellow | |
| C6H7N | |
| 500 mL | |
| 2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha | |
| CC1=CC=CC=N1 | |
| 93.129 | |
| CHEBI:50415 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 109-06-8 | |
| 93.129 | |
| BSKHPKMHTQYZBB-UHFFFAOYSA-N | |
| 7975 | |
| 2-methylpyridine |
| C6H7N | |
| MFCD00006332 | |
| 2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha | |
| CHEBI:50415 | |
| CC1=CC=CC=N1 |
Safety and Handling
EINECSNumber : (5)-0711
RTECSNumber : TJ4900000
TSCA : Yes