missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyridine 98.0+%, TCI America™

Catalog No. P041525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P041525ML 25 mL
P0415500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P041525ML Supplier TCI America Supplier No. P041525ML

Chemical Identifiers

CAS 109-06-8
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006332
InChI Key BSKHPKMHTQYZBB-UHFFFAOYSA-N
Synonym 2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha
PubChem CID 7975
ChEBI CHEBI:50415
IUPAC Name 2-methylpyridine
SMILES CC1=CC=CC=N1

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 128°C
Quantity 25 mL
UN Number 2313
Formula Weight 93.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methylpyridine
EINECSNumber (5)-0711
RTECSNumber TJ4900000
TSCA Yes