missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic Acid 97.0+%, TCI America™

Catalog No. p-7133411
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27601G Supplier TCI America Supplier No. M27601G

Chemical Identifiers

CAS 739364-95-5
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD19443280
InChI Key YHVGUXFUOSJCFJ-UHFFFAOYSA-N
PubChem CID 23091947
IUPAC Name 2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
SMILES CC1=NN2C=C(C=NC2=C1)C(=O)O

Specifications

Melting Point 238°C
Color White-Yellow
Quantity 1 g
Formula Weight 177.16
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic Acid