missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$73.16 - $73.16
Chemical Identifiers
| CAS | 16419-60-6 |
|---|---|
| Molecular Formula | C7H9BO2 |
| MDL Number | MFCD00093526 |
| InChI Key | NSJVYHOPHZMZPN-UHFFFAOYSA-N |
| PubChem CID | 2733267 |
| IUPAC Name | (2-methylphenyl)boronic acid |
Chemical Identifiers
| 16419-60-6 | |
| MFCD00093526 | |
| 2733267 |
| C7H9BO2 | |
| NSJVYHOPHZMZPN-UHFFFAOYSA-N | |
| (2-methylphenyl)boronic acid |
Specifications
| 16419-60-6 | |
| MFCD00093526 | |
| 2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl | |
| CC1=CC=CC=C1B(O)O | |
| 135.96 | |
| 135.96 | |
| 2-Methylphenylboronic Acid (contains varying amounts of Anhydride) |
| C7H9BO2 | |
| 5 g | |
| NSJVYHOPHZMZPN-UHFFFAOYSA-N | |
| (2-methylphenyl)boronic acid | |
| 2733267 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : ED7777777
TSCA : No