missing translation for 'onlineSavingsMsg'
Learn More

2-Methylphenethylamine, 97%

Catalog No. p-166458
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC351100010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC351100010 Supplier Thermo Scientific Chemicals Supplier No. 351100010

CAS: 55755-16-3 | C9H14N | 136.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55755-16-3
Molecular Formula C9H14N
Molecular Weight (g/mol) 136.22
MDL Number MFCD01310827
InChI Key OWOUKRYOZIZVFK-UHFFFAOYSA-O
Synonym 2-methylphenethylamine, 2-o-tolyl ethanamine, 2-2-methylphenyl ethylamine, benzeneethanamine, 2-methyl, 2-2-methylphenyl ethanamine, 2-o-tolyl ethylamine, 2-o-tolyl-ethylamine, 2-2-methylphenyl ethan-1-amine, 2-methyl-benzeneethanamine, +-tolylethylamine
PubChem CID 2063868
IUPAC Name 2-(2-methylphenyl)ethan-1-aminium
SMILES CC1=CC=CC=C1CC[NH3+]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.5340 to 1.5360 (20°C, 589nm)
Quantity 1 g
Beilstein 12, IV, 2638
Formula Weight 135.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methylphenethylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only