missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpentyl Acetate 98.0+%, TCI America™
Supplier: TCI America A1079500ML
Specifications
| 2-Methylpentyl Acetate | |
| Colorless | |
| C8H16O2 | |
| 500 mL | |
| Acetic Acid 2-Methylpentyl Ester | |
| CCCC(C)COC(C)=O | |
| 144.21 | |
| 144.21 | |
| Liquid |
| 7789-99-3 | |
| 160°C | |
| MFCD00060103 | |
| 3272 | |
| UZTVUHTZGGZFCI-UHFFFAOYNA-N | |
| 2-methylpentyl acetate | |
| 24625 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 7789-99-3 | |
| 144.21 | |
| UZTVUHTZGGZFCI-UHFFFAOYNA-N | |
| 24625 | |
| CCCC(C)COC(C)=O |
| C8H16O2 | |
| MFCD00060103 | |
| Acetic Acid 2-Methylpentyl Ester | |
| 2-methylpentyl acetate |
Safety and Handling
EINECSNumber : (2)-0734
RTECSNumber : AI9300000
TSCA : Yes